missing translation for 'onlineSavingsMsg'
Learn More

Gliclazide

Catalog No. AC461070010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461070010 1 g
AC461070050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461070010 Supplier Thermo Scientific Chemicals Supplier No. 461070010

CAS: 21187-98-4 | C15H21N3O3S | 323.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21187-98-4
Molecular Formula C15H21N3O3S
Molecular Weight (g/mol) 323.41
MDL Number MFCD00409893
InChI Key BOVGTQGAOIONJV-UHFFFAOYNA-N
Synonym gliclazide, glimicron, diamicron, nordialex, diaikron, gliclazida, gliklazid, glyclazide, gliclazidum inn-latin, gliclazida inn-spanish
PubChem CID 3475
ChEBI CHEBI:31654
IUPAC Name 1-(3,3a,4,5,6,6a-hexahydro-1H-cyclopenta[c]pyrrol-2-yl)-3-(4-methylphenyl)sulfonylurea
SMILES CC1=CC=C(C=C1)S(=O)(=O)NC(=O)NN1CC2CCCC2C1

Specifications

Melting Point 163.0°C to 169.0°C
Loss on Drying 0.5% max. (1 g, 105°C)
Infrared Spectrum Authentic
Quantity 1 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in dmf, slightly soluble in methanol, ethanol
Formula Weight 323.41
Percent Purity ≥98.5% (HPLC)
Chemical Name or Material Gliclazide
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Suspected of damaging fertility or the unborn child.
Precautionary Statement GHS P Statement Obtain special instructions before use. Wear protective gloves/protective clothing/eye protection/face protection. IF exposed or concerned: Get medical advice/attention.
EINECSNumber 244-260-5

RUO – Research Use Only