accessibility menu, dialog, popup

UserName

H3CSOOHNOHNN

CAS RN 21187-98-4

IUPAC Name: 1-(4-methylbenzenesulfonyl)-3-{octahydrocyclopenta[c]pyrrol-2-yl}urea
Synonyms: gliclazide diamicron MR genpharm brand of gliclazide novopharm brand of gliclazide helsinn brand of gliclazide gliclazide (product) novo-gliclazide glimicron gen gliclazide gliklazid
Molecular Weight (g/mol): 323.41
Molecular Formula: C15H21N3O3S
InChi Key: BOVGTQGAOIONJV-UHFFFAOYNA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)NC(=O)NN1CC2CCCC2C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Grade 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)