missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Glass Reagent Bottles with Hollow Interchangeable Hexagonal Glass Stoppers
Clear glass reagent bottles with interchangeable hollow glass stoppers are well suited for storing liquid solutions in the laboratory. Glass bottles and glass stoppers are autoclavable.
$25.32 - $197.29
Specifications
| Color | Clear |
|---|---|
| Description | Hollow stopper |
| For Use With (Application) | Solution Preparation, Solution Storage |
| Includes | Interchangeable hexagonal glass hollow stopper |
| Material | Borosilicate Glass |
Includes: Clear glass bottle, interchangeable hollow hexagonal glass stopper
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|
S39288
|
Eisco™
EISCH0162A |
30 mL |
Each for $25.32
|
|
|||||
|
S39290
|
Eisco™
CH0162C |
125 mL |
Each for $39.61
|
|
|||||
|
S39291
|
Eisco™
CH0162D |
250 mL |
Each for $44.54
|
|
|||||
|
S39292
|
Eisco™
CH0162E |
500 mL |
Each for $78.93
|
|
|||||
|
S39293
|
Eisco™
CH0162F |
1 L |
Each for $128.32
|
|
|||||
|
S39294
|
Eisco™
CH0162G |
2 L |
Each for $197.29
|
|
|||||
Description
- Made of high quality borosilicate 3.3 glass
- Narrow mouth with interchangeable hollow hexagonal glass stopper
- Bottle and stopper are autoclavable
Warranty
1 year limited manufacturer warranty
Specifications
| Clear | |
| Solution Preparation, Solution Storage | |
| Borosilicate Glass | |
| Non-sterile | |
| Yes | |
| No | |
| Cylindrical | |
| Liquids | |
| Glass | |
| Autoclavable |
| Hollow stopper | |
| Interchangeable hexagonal glass hollow stopper | |
| 1 | |
| Clear | |
| Reusable | |
| Narrow | |
| Reagent Bottle | |
| Unlabeled | |
| Stopper |