missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Glass Reagent Bottles with Hollow Interchangeable Hexagonal Glass Stoppers
Description
Includes
Clear glass bottle, interchangeable hollow hexagonal glass stopper
- Made of high quality borosilicate 3.3 glass
- Narrow mouth with interchangeable hollow hexagonal glass stopper
- Bottle and stopper are autoclavable
Warranty
1 year limited manufacturer warranty
Specifications
Specifications
| Capacity (Metric) | 2 L |
| Color | Clear |
| Description | Hollow stopper |
| Diameter (English) | 5 in. |
| Diameter (English) Inner Neck | 1.25 in |
| Diameter (Metric) Inner Neck | 31.75 mm |
| For Use With (Application) | Solution Preparation, Solution Storage |
| Includes | Interchangeable hexagonal glass hollow stopper |
| Material | Borosilicate Glass |
| Length (Metric) | 152.4mm |
| Show More |