missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Glass McCartney Bottles with Cap

Glass McCartney bottles with rubber lined aluminum threaded screw caps for storing liquid solutions

Specifications

Color Clear
For Use With (Application) Solution Storage
Includes Glass bottle, aluminum cap
Material Glass
Quantity 1 Ea.
View More Specs

Includes: Glass bottle, aluminum cap

Products 1
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
S39313
View Documents Promotion Details
Eisco™
CH0190A
28 mL N/A
Only null left
 
Description

Description

  • Includes glass bottle and aluminum cap
  • Cap is threaded and lined with rubber for a tight leak proof seal

Warranty

1 year limited manufacturer warranty

Specifications

Specifications

Clear
Glass bottle, aluminum cap
1 Ea.
Yes
No
Cylindrical
Unlabeled
Screw Cap
Solution Storage
Glass
Non-sterile
Reusable
Narrow
McCartney Bottle
Aluminum
Autoclavable
Videos
SDS
Documents

Documents