Learn More
Sartorius Glass Fiber Extraction Thimbles
Extraction thimbles been designed to be compatible with the Soxhlet apparatus, making them ideal for a variety of applications. Designed for versatility, the thimbles can be used with any extractor, including models like the Tectator, to facilitate efficient and high-yield extraction procedures.
Supplier: Sartorius FT1204025100
Description
- Composed of high-purity glass fiber for uncontaminated results
- Engineered for superior mechanical stability to withstand rigorous use
- Glass fiber thimbles offer remarkable resistance to high temperatures
- Designed for a precise fit across a diverse range of extractors
- Exhibits outstanding chemical resistance, ensuring reliability in various extraction processes
Chemical Identifiers
| Glass Fibers | |
| G400 | |
| Extraction Thimbles |
Specifications
| 25 mm | |
| 1.6 mm | |
| 100 mm |
| Glass Fibers | |
| G400 | |
| Extraction Thimbles |