Learn More
Gibco™ PBS, pH 7.2

Description
Gibco Phosphate-Buffered Saline (PBS), pH 7.2, is a balanced salt solution widely used in various cell culture applications. It is designed to maintain a stable pH and osmotic balance, making it ideal for washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.
Key features
- Without calcium and magnesium—the absence of calcium and magnesium is crucial for applications requiring the rinsing of chelators from the culture before cell dissociation
- Without phenol red—the lack of phenol red ensures that there is no interference with optical measurements and prevents potential effects on cell physiology
Applications
- Cell washing—ideal for washing cells before dissociation to remove any residual enzymes or chelators
- Sample transport—suitable for transporting cells or tissue samples while maintaining their viability
- Cell counting—can be used to dilute cells for accurate counting
- Reagent preparation—provides a stable environment for preparing various reagents used in cell culture
Why choose PBS vs. DPBS
- Simplicity—PBS is a simpler formulation compared to Dulbecco's Phosphate-Buffered Saline (DPBS), which includes additional components like potassium chloride and may be formulated with or without calcium, magnesium, glucose, and pyruvate. This simplicity makes PBS a versatile choice for general cell culture applications.
- Specific applications—PBS is preferred for applications where the presence of potassium or other additives in DPBS might interfere with experimental outcomes
Why choose pH 7.2 vs. pH 7.4 PBS
- Experimental requirements—the choice between pH 7.2 and pH 7.4 depends on the specific requirements of your cell culture system. pH 7.2 is often used for certain cell types and experimental conditions where a slightly more acidic environment is beneficial.
- Buffering capacity—pH 7.2 provides a slightly different buffering capacity, which can be crucial for maintaining the desired physiological conditions in specific experimental setups
Why choose Gibco PBS, pH 7.2
- Quality and consistency—Gibco products are manufactured in cGMP, ISO-certified facilities, ensuring the highest quality and consistency. Each batch undergoes rigorous testing for osmolality, pH levels, sterility, and the presence of endotoxins.
- Track record—Gibco cell culture products have a long-standing reputation for excellence in the field of cell culture, with numerous publications citing the use of Gibco products. This track record of reliability and performance makes Gibco PBS the preferred choice for researchers worldwide.
Gibco PBS, pH 7.2 is a reliable and consistent solution for a wide range of applications. Choose Gibco products for your cell culture needs and benefit from our decades of expertise and innovation in the field.
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Chemical Name or Material | Phosphate Buffered Saline (PBS) |
| Color | White |
| Concentration | 1 X |
| For Use With (Application) | Mammalian Cell Culture |
| pH | 7.2 |
| Physical Form | Liquid |
| Product Line | Gibco |
| Quantity | 500 mL |
| Solution Type | Phosphate Buffered Saline |
| Sterility | Sterile-filtered |
| Show More |
Frequently Asked Questions (FAQs)
Our PBS, pH 7.2 and PBS, pH 7.4 products are tested for endotoxin and the endotoxin levels are shown on the Certificate of Analysis (CoA) for the particular lot number of the product.
We offer PBS - Phosphate-Buffered Saline (10X) pH 7.4, RNase-free (Cat. Nos. AM9624 and AM9625), which is molecular biology grade and is certified to be RNase-free. However, it is not sterile and hence is not cell culture-grade.
All our other PBS products (Cat. Nos. 70011xxx, 10010xxx, 70013xxx, 20012xxx) are filter-sterilized using a 0.1 µm filter and are cell culture grade (ready to use directly on cells). However, they are not tested for nuclease activity.
We offer PBS - Phosphate-Buffered Saline (10X) pH 7.4, RNase-free (Cat. Nos. AM9624 and AM9625) that is molecular biology grade and is certified to be RNase-free. Our other PBS products are made with distilled water and are filter-sterilized, but are not tested for RNase and DNase activity.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.