Learn More
Gibco™ PBS (10X), pH 7.4

Description
Phosphate-buffered saline (PBS) is a balanced salt solution that is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. PBS is formulated without calcium and magnesium for rinsing chelators from the culture before cell dissociation. Concentrated forms of Gibco™ PBS may require pH adjustment at the time of dilution (see protocol for details).
We offer a variety of Gibco™ PBS formulations for a range of cell culture applications.
This PBS is manufactured as follows:
Without: Calcium, Magnesium, Phenol Red
The complete formulation is available.
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Chemical Name or Material | Phosphate Buffered Saline (PBS) |
| Color | White |
| Concentration | 10 X |
| For Use With (Application) | Mammalian Cell Culture |
| pH | 7.4 |
| Physical Form | Liquid |
| Product Line | Gibco |
| Quantity | 500 mL |
| Solution Type | Phosphate Buffered Saline |
| Sterility | Sterile-filtered |
| Show More |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.