missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ PBS (10X), pH 7.4
GREENER_CHOICE

Catalog No. 70011069
Encompass
Change view
Click to view available options
Quantity:
500 mL
10 x 500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70011069 10 x 500 mL
70011044 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 70011069 Supplier Gibco™ Supplier No. 70011069
Only null left

Phosphate-buffered saline (PBS) is a balanced salt solution that is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.

Phosphate-buffered saline (PBS) is a balanced salt solution that is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. PBS is formulated without calcium and magnesium for rinsing chelators from the culture before cell dissociation. Concentrated forms of Gibco™ PBS may require pH adjustment at the time of dilution (see protocol for details).

We offer a variety of Gibco™ PBS formulations for a range of cell culture applications.

This PBS is manufactured as follows:

Without: Calcium, Magnesium, Phenol Red

The complete formulation is available.

Order Info

Shipping Condition: Room Temperature

Specifications

Chemical Name or Material Phosphate Buffered Saline (PBS)
Color White
Concentration 10 X
For Use With (Application) Mammalian Cell Culture
pH 7.4
Physical Form Liquid
Product Line Gibco
Quantity 10 x 500 mL
Solution Type Phosphate Buffered Saline
Sterility Sterile-filtered
Includes No Sodium Pyruvate
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Osmolality 2800 - 3100 mOsm/kg
Show More Show Less
Are your PBS products tested for endotoxin levels?

Our PBS, pH 7.2 and PBS, pH 7.4 products are tested for endotoxin and the endotoxin levels are shown on the Certificate of Analysis (CoA) for the particular lot number of the product.

Are your PBS products sterile and suitable for cell culture?

We offer PBS - Phosphate-Buffered Saline (10X) pH 7.4, RNase-free (Cat. Nos. AM9624 and AM9625), which is molecular biology grade and is certified to be RNase-free. However, it is not sterile and hence is not cell culture-grade.

All our other PBS products (Cat. Nos. 70011xxx, 10010xxx, 70013xxx, 20012xxx) are filter-sterilized using a 0.1 µm filter and are cell culture grade (ready to use directly on cells). However, they are not tested for nuclease activity.

Are your PBS products DNase- and RNase-free?

We offer PBS - Phosphate-Buffered Saline (10X) pH 7.4, RNase-free (Cat. Nos. AM9624 and AM9625) that is molecular biology grade and is certified to be RNase-free. Our other PBS products are made with distilled water and are filter-sterilized, but are not tested for RNase and DNase activity.

Is a precipitate in PBS normal?

No, this is not normal and if there is a precipitate in the product, it cannot be used. If the precipitate will not go back into solution, please contact Technical Support at techsupport@thermofisher.com for a replacement.

What is the difference between PBS and DPBS?

PBS has no potassium chloride (KCl) and it has a slightly lower level of phosphate (PO4) than DPBS. PBS is used for washing cells when viability isn't critical.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.