missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ MEM, GlutaMAX™ Supplement DFS Item

Minimum Essential Medium (MEM) is one of the most commonly used of all cell culture media.

$96.00 - $812.00

Specifications

Cell Line HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, and fibroblasts
Cell Type Primary Rat Astrocytes
Classification Animal Origin-free
Concentration 1 X
Form Liquid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
41090036 DFS Item
View Documents
Gibco™
41090036
500 mL
Each for $96.00
Only null left
 
41090101 DFS Item
View Documents
Gibco™
41090101
10 x 500 mL
Each for $812.00
Only null left
 
Description

Description

Minimum Essential Medium (MEM) is one of the most commonly used of all cell culture media. MEM can be used with a variety of suspension and adherent mammalian cells, including HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, fibroblasts, and primary rat astrocytes. We offer a variety of Gibco™ MEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This MEM is modified as follows:

With: GlutaMAX™, Phenol Red

Without: HEPES

The complete formulation is available.

Gibco™ MEM, developed by Harry Eagle, was based on his earlier formulation of Basal Medium Eagle (BME). Many other modifications of MEM followed, including Glasgow's MEM, MEM α, DMEM, and Temin's Modification. MEM is available with Earle's salts for use in a CO2 incubator, or with Hanks' salts for use without CO2. Gibco™ MEM with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format. This product is made with Earle's salts.

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, and fibroblasts
Animal Origin-free
Liquid
Standard Serum Supplementation
Sterile-filtered
No HEPES, No Sodium Pyruvate
Gibco, GlutaMAX
Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Primary Rat Astrocytes
1 X
MEM (Minimum Essential Medium)
Sterile-filtered
Low Glucose, GlutaMAX, Phenol Red
cGMP-compliant under the ISO 13485 standard
Room Temperature
Videos
SDS
Documents

Documents

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.