Learn More
Gibco™ HEPES (1 M)

Description
Gibco HEPES (4-(2-hydroxyethyl)-1-piperazineethanesulfonic acid) is a zwitterionic organic chemical buffering agent commonly used in cell culture media. The addition of 10–25 mM HEPES provides extra buffering capacity when cell culture requires extended periods of manipulation outside of a CO2 incubator.
Gibco HEPES (N-2-hydroxyethylpiperazine-N-2-ethane sulfonic acid) is a good buffering choice for many cell culture systems because it:
- Is membrane impermeable
- Has limited effect on biochemical reactions
- Is chemically and enzymatically stable
- Has a very low visible and UV light absorbance
cGMP manufacturing and quality system
For supply chain continuity, we manufacture this product at two separate facilities located in Grand Island, NY, and Scotland, UK. Both sites are compliant with cGMP manufacturing requirements and are certified to the ISO 13485 standard.
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Chemical Name or Material | Zwitterionic Organic Chemical Buffer |
| CAS | 7365-45-9 |
| Concentration | 1 M |
| For Use With (Application) | Chromatin Biology |
| pH | 7 to 7.5 |
| Physical Form | Liquid |
| Quantity | 20 x 100 mL |
| Solution Type | Buffer |
| Sterility | Sterile |
| Sterilization Method | Sterile-filtered |
| Show More |
Frequently Asked Questions (FAQs)
This product should be stored at 2-8 degrees C and should not be frozen. The biggest issue with accidentally freezing this product is its solubility. If this product was accidentally frozen, we recommend placing it in a 2-8 degree environment and allowing it to slowly thaw overnight. If the product is fully thawed and shows no signs of precipitation, then it should still be usable, but we cannot guarantee effectiveness.
1M HEPES is in water with a pH range of 7.2 - 7.5.
Sorry, we do not test endotoxin levels for this product.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.