missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ HBSS, calcium, magnesium, no phenol red
GREENER_CHOICE

Catalog No. 14025092
Encompass
Change view
Click to view available options
Quantity:
500 mL
1000 mL
10 x 500 mL
6 x 1000 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14025092 500 mL
14025134 10 x 500 mL
14025076 1000 mL
14025126 6 x 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14025092 Supplier Gibco™ Supplier No. 14025092

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.

Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media, or for reagent preparation.

We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications.

This HBSS is modified as follows:

With: Calcium, Magnesium, Glucose

Without: Phenol Red, Sodium Pyruvate

The complete formulation is available.

Order Info

Shipping Condition: Room Temperature

Specifications

Chemical Name or Material Hanks' Balanced Salt Solution (HBSS)
Color Clear
Concentration 1X
For Use With (Application) Mammalian Cell Culture
pH 6.7 to 7.8
Physical Form Liquid
Product Line Gibco
Quantity 500 mL
Solution Type pH Buffer, Storage Solution and Cleaning Solution
Sterility Sterile
Includes No Sodium Pyruvate
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Sterilization Method Sterile-filtered
Osmolality 270 to 305 mOsm/kg
Grade Cell Culture
Show More Show Less
What is the pH of HBSS, calcium, magnesium, no phenol red at room temperature.

At room temperature, HBSS, calcium, magnesium, no phenol red has a pH of 6.7 - 7.8.

Can I store Hank's Balanced Salt Solution (HBSS) in the refrigerator?

The recommended storage condition for our HBSS products is at 15-30 degrees C. Generally, storage at 4 degrees C will not impact these products. The only concern would be for the 10x HBSS solutions with calcium and magnesium as these salts might precipitate when stored outside the recommended temperature ranges. As long as you don't see any precipitation upon refrigeration, the product will be fine.

Do I need to add additional HEPES to Hank's Balanced Salt Solution (HBSS) (Cat. No. 14025xxx)?

HBSS (Cat. No. 14025xxx) does not contain HEPES and it needs to be externally supplemented. The complete formulation is available here (https://www.thermofisher.com/us/en/home/technical-resources/media-formulation.153.html).

I understand that some media are worse than others for fluorescence imaging. How do I choose?

Most media contain phenol red, which can quench fluorescent dyes in the visible wavelengths. Most media also contain autofluorescent components, such as riboflavin, which can reduce signal-to-background. We offer FluoroBrite DMEM and HEPES-based Live Cell Imaging Solution, which have been optimized for fluorescent imaging. We also offer a number of media without phenol red. But if none of these are reasonable options for your experiment, then we also offer BackDrop Background Suppressor ReadyProbes Reagent, which can be added to quench media autofluorescence.

Should I be concerned about phenol red in my media when labeling my live cells with fluorescent dyes?

Some cell types accumulate phenol red, and this can pose a problem in the use of many fluorescent probes. Phenol red can quench visible-wavelength dyes and, although phenol red is non-fluorescent, various impurities may be fluorescent. We have many phenol red-free media to choose from. Our Live Cell Imaging Solution (HEPES-based) and our FluoroBrite DMEM have been optimized to be phenol red-free as well as to be non-autofluorescent.

Why do you offer HBSS with and without phenol red?

Phenol red is a pH indicator which allows you to determine the optimal atmospheric conditions for your cell line during your particular assays. We offer both versions to allow you the option to utilize this indicator in your media.

Can I use HBSS (with calcium and magnesium) to wash my cells before dissociation?

No, a saline solution that contains calcium and magnesium should not be used to wash cells before dissociation.

Is a precipitate in HBSS (with calcium and magnesium) normal?

No, this is not normal and if there is a precipitate in the product, it cannot be used. If the precipitate will not go back into solution, please contact Technical Support at techsupport@thermofisher.com for a replacement.

Do you offer media without phenol red (phenol red-free media)?

Yes, please visit this page for a list of our phenol red-free media (http://tools.thermofisher.com/content/sfs/brochures/Phenol_RedFree_Media_Glance.pdf).


For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.