Learn More
Gibco™ DPBS, calcium, magnesium, glucose, pyruvate
Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents.
$76.50 - $76.50
Specifications
| Chemical Name or Material | DPBS |
|---|---|
| Concentration | 1 X |
| For Use With (Application) | Mammalian Cell Culture |
| Physical Form | Liquid |
| Product Line | Gibco |
Description
Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.
This DPBS is modified as follows:
With: Calcium, Magnesium, Glucose, Sodium Pyruvate
Without: Phenol Red
The complete formulation is available.
Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.
Order Info
Shipping Condition: Room Temperature
Specifications
| DPBS | |
| Mammalian Cell Culture | |
| Gibco | |
| Sterile-filtered | |
| cGMP-compliant under the ISO 13485 standard |
| 1 X | |
| Liquid | |
| Dulbecco's Phosphate Buffered Saline | |
| Sodium Pyruvate |
For Research Use Only. Not for use in diagnostic procedures.