Learn More
Gibco™ DPBS, calcium, magnesium

Description
Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications.
This DPBS is modified as follows:
With: Calcium, Magnesium
Without: Phenol Red
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Chemical Name or Material | DPBS |
| Concentration | 1X |
| For Use With (Application) | Washing and Staining Cells |
| pH | 7.0 to 7.2 |
| Physical Form | Liquid |
| Product Line | Gibco |
| Quantity | 10 x 500 mL |
| Solution Type | Cleaning, Storage, pH Buffer |
| Sterility | Sterile |
| Includes | No Sodium Pyruvate |
| Show More |
Frequently Asked Questions (FAQs)
Sorry, we do not test the conductivity of our DPBS products and hence cannot provide a value or range.
Our DPBS products are for cell culture use and have not been tested for DNase/RNase activity. We do not recommend using these products for RNA work.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.