missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ DPBS, calcium, magnesium
GREENER_CHOICE

Catalog No. 14040133
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
10 x 500 mL
6 x 1000 mL
Osmolality:
270 to 300 mOsm/kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Osmolality
14040133 500 mL 270 to 300 mOsm/kg
14040182 10 x 500 mL 270 to 300 mOsm/kg
14040141 100 mL 270 to 300 mOsm/kg
14040117 1000 mL 270 to 300 mOsm/kg
14040216 6 x 1000 mL 270 to 300 mOsm/kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14040133 Supplier Gibco™ Supplier No. 14040133

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.

Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications.

This DPBS is modified as follows:

With: Calcium, Magnesium

Without: Phenol Red

Order Info

Shipping Condition: Room Temperature

Specifications

Chemical Name or Material DPBS
Concentration 1X
For Use With (Application) Washing and Staining Cells
pH 7.0 to 7.2
Physical Form Liquid
Product Line Gibco
Quantity 500 mL
Solution Type Cleaning, Storage, pH Buffer
Sterility Sterile
Includes No Sodium Pyruvate
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Sterilization Method Sterile-filtered
Osmolality 270 to 300 mOsm/kg
Grade Cell Culture
Show More Show Less
Can I get a specific value for the conductivity for your DPBS products?

Sorry, we do not test the conductivity of our DPBS products and hence cannot provide a value or range.

Are your DPBS products DNase/RNase-free? Can I use them for RNA work?

Our DPBS products are for cell culture use and have not been tested for DNase/RNase activity. We do not recommend using these products for RNA work.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.