missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CD Hybridoma AGT™ Medium DFS Item

Animal origin-free, chemically defined, protein-free medium

Supplier:  Gibco™ 12372017

 View more versions of this product

Catalog No. 12372017


Only null left

Description

Description

Gibco CD Hybridoma Medium is specifically developed for the ability to support the growth of hybridomas for antibody production.

  • Ability to support suspension and stationary hybridomas and myelomas
  • Formulated without L-glutamine for stability
  • Animal origin-free, protein-free and chemically defined formulation
  • Convenient, easy-to-use Advanced Granulation Technology (AGT™) format

Ability to support suspension and stationary hybridomas and myelomas

  • Suitable for the culture of recombinant myeloma lines as well as traditional hybridomas, Gibco CD Hybridoma Medium provides high cell densities and IgG production
  • Cholesterol-dependent cultures such as NS0 require the addition of 250X Cholesterol Lipid Concentrate
Formulated without L-glutamine for stability
  • Gibco CD Hybridoma Medium requires the addition of 8 mM L-glutamine or GlutaMAX™ supplement before use
  • GlutaMAX supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format
Animal origin-free, protein-free and chemically defined formulation
  • Gibco CD Hybridoma Medium is animal origin-free, protein-free and chemically defined, allowing for easier purification of your protein of interest
  • Gibco chemically defined media contain no proteins, hydrolysates or components of unknown composition
Convenient, easy-to-use Advanced Granulation Technology (AGT™) format
  • The AGT format is an easy-to-use dry powder format. The granules dissolve instantly and pH-adjusts for fast media preparation
  • Complete reconstitution instructions are available in the product manual

Antibody and Cell Culture Production, Bioproduction, Cell Culture, Mammalian Cell Culture

Specifications

Specifications

Myeloma Cells, Hybridomas
Powder (AGT)
Serum-free
Sodium Pyruvate, Sodium Bicarbonate
AGT, Gibco
Wet Ice
Storage conditions: 2-8°C, dark and dry
Shipping conditions: Gel packs
Shelf life: 24 months from date of manufacture
Animal Origin-free, Chemically-defined, Protein-free
Hybridoma Medium
Non-sterile
No Glutamine, No Phenol Red
10 L
≤10 EU/mL, ≤ 10 EU/ml
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.