missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CD Hybridoma AGT™ Medium

Catalog No. p-4924966
Encompass
Change view
Click to view available options
Quantity:
1 L
10 L
100 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12372025 1 L
12372017 10 L
12372001 100 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12372025 Supplier Gibco™ Supplier No. 12372025
Only null left

Animal origin-free, chemically defined, protein-free medium

Gibco CD Hybridoma Medium is specifically developed for the ability to support the growth of hybridomas for antibody production.

  • Ability to support suspension and stationary hybridomas and myelomas
  • Formulated without L-glutamine for stability
  • Animal origin-free, protein-free and chemically defined formulation
  • Convenient, easy-to-use Advanced Granulation Technology (AGT™) format

Ability to support suspension and stationary hybridomas and myelomas

  • Suitable for the culture of recombinant myeloma lines as well as traditional hybridomas, Gibco CD Hybridoma Medium provides high cell densities and IgG production
  • Cholesterol-dependent cultures such as NS0 require the addition of 250X Cholesterol Lipid Concentrate
Formulated without L-glutamine for stability
  • Gibco CD Hybridoma Medium requires the addition of 8 mM L-glutamine or GlutaMAX™ supplement before use
  • GlutaMAX supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format
Animal origin-free, protein-free and chemically defined formulation
  • Gibco CD Hybridoma Medium is animal origin-free, protein-free and chemically defined, allowing for easier purification of your protein of interest
  • Gibco chemically defined media contain no proteins, hydrolysates or components of unknown composition
Convenient, easy-to-use Advanced Granulation Technology (AGT™) format
  • The AGT format is an easy-to-use dry powder format. The granules dissolve instantly and pH-adjusts for fast media preparation
  • Complete reconstitution instructions are available in the product manual

Antibody and Cell Culture Production, Bioproduction, Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Wet ice

Specifications

Cell Type Myeloma Cells, Hybridomas
Classification Animal Origin-free, Chemically-defined, Protein-free
Form Powder (AGT)
Product Type Hybridoma Medium
Serum Level Serum-free
Sterility Non-sterile
With Additives Sodium Pyruvate, Sodium Bicarbonate
Without Additives No Glutamine, No Phenol Red
Product Line AGT, Gibco
Quantity 1 L
Shipping Condition Wet Ice
Endotoxin Level ≤10 EU/mL, ≤ 10 EU/ml
Content And Storage Storage conditions: 2-8°C, dark and dry
Shipping conditions: Gel packs
Shelf life: 24 months from date of manufacture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.