missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ AmnioMAX™ C-100 Basal Medium
GREENER_CHOICE

Part of kit used to prepare fully-supplemented Gibco AmnioMAX C-100 Complete Medium developed for short term culture of human amniotic fluid cells for cytogenetic studies and in vitro diagnostic procedures

$61.00 - $229.00

Specifications

Form Liquid
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
17001082
GREENER_CHOICE
Encompass
View Documents Promotion Details
Gibco™
17001082
90 mL
Each for $61.00
Only null left
 
17001074
GREENER_CHOICE
Encompass
View Documents Promotion Details
Gibco™
17001074
450 mL
Each for $229.00
Only null left
 
Description

Description

Gibco AmnioMAX C-100 Complete Medium is two-part kit for preparation of a complete medium that requires no additional supplementation.

  • Quality and performance testing
  • Every lot of Gibco AmnioMAX C-100 Complete Medium is performance tested by a certified US reference cytogenetics laboratory to ensure consistently superior performance
  • Pooled primary human amniotic fluid samples are cultured for six days in Gibco AmnioMAX C-100 Complete Medium before measuring total number of colonies and total number of mitotic colonies
  • In addition, each lot is tested for sterility, pH and osmolality
  • Unique, optimized formulation
  • Gibco AmnioMAX C-100 Complete Medium contains Fetal Bovine Serum (FBS), gentamicin, and L-glutamine to maximize cell attachment and growth
  • It has enhanced buffering system that provides greater pH stability during culture manipulations
  • Sustainable packaging

Amniotic Fluid and Chorionic Villus Culture, Cell Analysis, Cell Culture, Cell Proliferation, Cell Viability, Proliferation and Function, Clinical, Clinical and Translational Research, Cytogenetics, Mammalian Cell Culture

Order Info

Shipping Condition: Room Temperature

Compliance

Gibco AmnioMAXC-100 Basal Medium is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

Specifications

Specifications

Liquid
Sterile-filtered
Sterile-filtered
Videos
Safety and Handling

Safety and Handling

ShelfLife : 16 Months From Date of Manufacture

SDS
Documents

Documents

Product Intended Use: For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.