Learn More
Gibco™ AmnioMAX™ C-100 Basal Medium

Description
Gibco AmnioMAX C-100 Complete Medium is two-part kit for preparation of a complete medium that requires no additional supplementation.
- Quality and performance testing
- Every lot of Gibco AmnioMAX C-100 Complete Medium is performance tested by a certified US reference cytogenetics laboratory to ensure consistently superior performance
- Pooled primary human amniotic fluid samples are cultured for six days in Gibco AmnioMAX C-100 Complete Medium before measuring total number of colonies and total number of mitotic colonies
- In addition, each lot is tested for sterility, pH and osmolality
- Unique, optimized formulation
- Gibco AmnioMAX C-100 Complete Medium contains Fetal Bovine Serum (FBS), gentamicin, and L-glutamine to maximize cell attachment and growth
- It has enhanced buffering system that provides greater pH stability during culture manipulations
- Sustainable packaging
Amniotic Fluid and Chorionic Villus Culture, Cell Analysis, Cell Culture, Cell Proliferation, Cell Viability, Proliferation and Function, Clinical, Clinical and Translational Research, Cytogenetics, Mammalian Cell Culture
Order Info
Shipping Condition: Room Temperature
Compliance
Gibco AmnioMAXC-100 Basal Medium is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
Specifications
Specifications
| Cell Type | Amniocyte Cell |
| Form | Liquid |
| Product Type | Amniotic Fluid Cell Basal Medium |
| Serum Level | Standard Serum Supplementation |
| Sterility | Sterile-filtered |
| Sterilization Method | Sterile-filtered |
| With Additives | Glutamine, HEPES |
| Product Line | AmnioMAX |
| Quantity | 450 mL |
| Shipping Condition | Room Temperature |
| Show More |
Product Intended Use: For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.