missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GFP-STAT1

Catalog No. PV5211
Encompass
Change view
Click to view available options
Quantity:
10 nmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV5211 10 nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PV5211 Supplier Invitrogen™ Supplier No. PV5211

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

GFP-STAT1

STAT1 expressed as a fusion with GFP (Green Fluorescent Protein) to act as a physiological substrate for the Janus family of tyrosine kinases.

For use in combination with the LanthaScreen™ Tb-anti-STAT1 [pTyr701] Antibody (PV4844) to evaluate inhibitors in medium to high throughput screening applications.

Specifications

Assay Entry Biochemical terbium phospho-specific activity
Content And Storage GFP-STAT1 protein substrate in 50 mM Tris-HCl pH 7.5, 100 mM NaCl, 10% Glycerol, 1 mM DTT. Store at -80°C.
Detection Method Fluorescence
Quantity 10 nmol
Substrate STAT1
Substrate Properties Protein-Based Substrate
Substrate Type Kinase Substrate
Target Enzyme Tyrosine Kinases
Label or Dye GFP (EmGFP)
Molecular Weight (g/mol) 116000 Da
Shipping Condition Dry Ice
Target Entry JAK1, JAK2 JH1 JH2, JAK2, TYK2, JAK3, JAK2 JH1 JH2 V617F
Validated Kinases JAK1, JAK2, JAK3, TYK2
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.