missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GFP-C-Jun (1-179)

Catalog No. PV4446
Encompass
Change view
Click to view available options
Quantity:
20 nmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV4446 20 nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PV4446 Supplier Thermo Scientific™ Supplier No. PV4446

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A truncated version (residues 1-179) of c-Jun has been expressed as a fusion with GFP (Green Fluorescent Protein) to act as a substrate for the JNK family of kinases.

A truncated version (residues 1-179) of c-Jun has been expressed as a fusion with GFP (Green Fluorescent Protein) to act as a substrate for the JNK family of kinases.

For use in combination with the LanthaScreen™ Tb-anti-c-Jun [pSer73] antibody (PV4453) to evaluate inhibitors in medium to high throughput screening applications.

Specifications

Assay Entry Biochemical terbium phospho-specific activity
Content And Storage GFP-c-Jun protein substrate in 10 mM Tris, pH 7.5, 150 mM NaCl, 10% glycerol, 1 mM DTT. Store (at -80°C.)
Detection Method Fluorescence
Quantity 20 nmol
Substrate c-Jun (1-179)
Substrate Properties Protein-Based Substrate
Substrate Type Kinase Substrate
Target Enzyme Serine/Threonine Kinases
Label or Dye GFP (EmGFP)
Molecular Weight (g/mol) 51.6 kDa
Shipping Condition Dry Ice
Target Entry NLK
Validated Kinases MAPK8, MAPK9, MAPK10, NLK
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.