missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GFP-ATF2 (19-96)

Catalog No. PV4445
Encompass
Change view
Click to view available options
Quantity:
20 nmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV4445 20 nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PV4445 Supplier Invitrogen™ Supplier No. PV4445

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

GFP-ATF2 (19-96)

A truncated version (residues 19-96) of ATF2 has been expressed as a fusion with GFP (Green Fluorescent Protein) to act as a substrate for the JNK and p38 families of kinases.

For use in combination with the LanthaScreen™ Tb-anti-ATF2 [pThr71] antibody (PV4451) to evaluate inhibitors in medium to high throughput screening applications.

Specifications

Assay Entry Biochemical terbium phospho-specific activity
Content And Storage GFP-ATF2 protein substrate in 10 mM Tris, pH 7.5, 150 mM NaCl, 10% glycerol, 1mM DTT. Store at -80°C.
Detection Method Fluorescence
Form Liquid
Quantity 20 nmol
Substrate ATF2 (19-96)
Substrate Properties Protein-Based Substrate
Substrate Type Kinase Substrate
Target Enzyme Serine/Threonine Kinases
Label or Dye GFP (EmGFP)
Molecular Weight (g/mol) 478000 Da
Shipping Condition Dry Ice
Target Entry MAPK13, MAPK8, MAPK9, MAPK14, MAPK3, MAPK1, MAPK11, MAPK12, MAPK10
Validated Kinases MAPK1, MAPK3, MAPK8, MAPK9, MAPK10, MAPK11, MAPK12, MAPK13, MAPK14
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.