missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GFP-4E-BP1

Catalog No. PV4759
Encompass
Change view
Click to view available options
Quantity:
20 nmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV4759 20 nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PV4759 Supplier Thermo Scientific™ Supplier No. PV4759

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

4E-BP1 expressed as a fusion with GFP (Green Fluorescent Protein) to act as a physiological substrate for FRAP1 (mTor).

4E-BP1 expressed as a fusion with GFP (Green Fluorescent Protein) to act as a physiological substrate for FRAP1 (mTor).

For use in combination with the LanthaScreen™ Tb-anti-4E-BP1 [pThr46] Antibody (Cat. No. PV4757) to evaluate inhibitors in medium to high throughput screening applications. An optimized mTor assay buffer (PV4794) is also available.

Specifications

Assay Entry Biochemical terbium phospho-specific activity
Content And Storage GFP-4E-BP1 protein substrate in 10 mM Tris, pH 7.5, 150 mM NaCl, 10% glycerol, 2 mM DTT

Store at -80°C.
Detection Method Fluorescence
Form Liquid
For Use With (Application) Pharma and biopharma, protein assays and analysis, target and lead identification and validation
Quantity 20 nmol
Substrate 4E-BP1
Substrate Properties Protein-Based Substrate
Substrate Type Kinase Substrate
Target Enzyme Serine/Threonine Kinases
Label or Dye GFP (EmGFP)
Molecular Weight (g/mol) 45,600 Da
Packaging 1 Each
Product Type GFP-4E-BP1
Shipping Condition Dry Ice
Storage Requirements –80°C
Target Entry mTOR, FRAP1
Validated Kinases FRAP1 (mTOR), FRAP1 (mTOR)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.