missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Germanium Standard For ICP, 1000μg/mL in H2O, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC PLGE92X
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Ge | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| Germanium Standard | |
| Assurance Certificate of Analysis reports up to 39 elements at ppb levels. | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| MFCD00085310 | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| GNPVGFCGXDBREM-UHFFFAOYSA-N | |
| Water/Trace F- | |
| 72.63 | |
| 6326954 | |
| Assurance |
| Triple-leached LDPE bottle | |
| 7440-56-4 , 7732-18-5 | |
| Ge | |
| AA, ICP | |
| Certified Reference Material | |
| [Ge] | |
| germanium | |
| 500 mL | |
| CHEBI:30441 | |
| Matrix Concentration Cat. No. H2O/tr. HF 1000μg/mL PLGE9-2Y, -2X 10,000 PLGE9-3Y, -3X |
Chemical Identifiers
| 7440-56-4 | |
| 72.63 | |
| GNPVGFCGXDBREM-UHFFFAOYSA-N | |
| CHEBI:30441 | |
| [Ge] |
| Ge | |
| MFCD00085310 | |
| 6326954 | |
| germanium |
Safety and Handling
DOTInformation : Nonhazardous
Recommended Storage : Ambient
Product intended for laboratory use only