missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Gentiopicroside

Catalog No. AC466830010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466830010 1 g
AC466832500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466830010 Supplier Thermo Scientific Chemicals Supplier No. 466830010

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20831-76-9
Molecular Formula C16H20O9

Specifications

Quantity 1 g
Melting Point 191.0°C
Formula Weight 356.33
InChI Key DUAGQYUORDTXOR-GPQRQXLASA-N
SMILES OC[C@H]1O[C@@H](O[C@@H]2OC=C3C(=O)OCC=C3[C@H]2C=C)[C@H](O)[C@@H](O)[C@@H]1O
IUPAC Name (5R,6S)-5-ethenyl-6-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-1H,3H,5H,6H-pyrano[3,4-c]pyran-1-one
Molecular Weight (g/mol) 356.33
Chemical Name or Material Gentiopicroside
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

EINECSNumber 244-070-2

RUO – Research Use Only