missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gentian Violet, 2% (w/v), Ricca Chemical
Specifications
Specifications
| CAS Min % | 96.0, 1.92 |
| CAS Max % | 100.0, 2.0 |
| Packaging | Natural Poly Bottle |
| Solvent or Matrix | Aqueous Solution |
| Color | Violet |
| Quantity | 120 mL |
| Chemical Name or Material | Gentian Violet |