missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ricca Chemical Company Gentian Violet, 1% (w/v), Ricca Chemical
Supplier: Ricca Chemical Company 32304
Specifications
548-62-9,7732-18-5 | |
100.0,0.71 | |
ZXJXZNDDNMQXFV-UHFFFAOYSA-M | |
[4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride | |
11057 | |
Natural Poly Bottle |
97.0,0.68 | |
C25H30ClN3 | |
CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-] | |
407.986 | |
CHEBI:41688 | |
120 mL |
Chemical Identifiers
548-62-9 | |
407.986 | |
11057 | |
[4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride |
C25H30ClN3 | |
ZXJXZNDDNMQXFV-UHFFFAOYSA-M | |
CHEBI:41688 | |
CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-] |