missing translation for 'onlineSavingsMsg'
Learn More

Gentian Violet, 1% Aqueous, Certified, LabChem™

Supplier:  LabChem LC148101

Quantity 500 mL
Packaging Poly Bottle

 View more versions of this product

Catalog No. LC148101

Only null left

Description

Description

Specifications

Chemical Identifiers

548-62-9 , 7732-18-5
407.986
11057
[4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
C25H30ClN3
ZXJXZNDDNMQXFV-UHFFFAOYSA-M
CHEBI:41688
CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]

Specifications

Gentian Violet
548-62-9 , 7732-18-5
C25H30ClN3
Soluble in water
ZXJXZNDDNMQXFV-UHFFFAOYSA-M
[4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
11057
407.99
Poly Bottle
Purple
Liquid
1% Aqueous
99, 1
C25H30CIN3
Passes Test
CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
407.986
CHEBI:41688
Certified
Carbon monoxide; Carbon dioxide; Hydrogen chloride; Nitrogen oxides
500 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Suspected of causing cancer.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
f exposed or concerned: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Warning

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions