missing translation for 'onlineSavingsMsg'
Learn More

Gentamycin Sulfate, USP, MP Biomedicals™

$596.36 - $996.74

Chemical Identifiers

CAS 1405-41-0
Synonym Gentamycin Sulfate
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ICN19005705
View Documents Promotion Details
N/A 5 g
Each for $596.36
Only null left
 
ICN19005710
View Documents Promotion Details
N/A 10 g
Each for $996.74
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1405-41-0
Gentamycin Sulfate

Specifications

Gentamicin Sulfate
1405-41-0
Gentamycin Sulfate
Powder
White
Gentamycin Sulfate, USP
Videos
SDS
Documents

Documents