missing translation for 'onlineSavingsMsg'
Learn More

Gentamycin Sulfate, USP, MP Biomedicals™

Catalog No. ICN19005705
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19005705 5 g
ICN19005710 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19005705 Supplier MP Biomedicals Inc Supplier No. 0219005705

Chemical Identifiers

CAS 1405-41-0
Synonym Gentamycin Sulfate

Specifications

Type Gentamicin Sulfate
Physical Form Powder
Color White
Quantity 5 g
Chemical Name or Material Gentamycin Sulfate, USP