missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Gentamycin Sulfate

CAS: 1405-41-0 | C21H43N5O7·H2SO4

Supplier:  Thermo Scientific Chemicals 613980010

Catalog No. AC613980010


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Gentamicin Sulfate
1405-41-0
1 g
MFCD00270181
Garamycin, Gentamicin Sulfate
Gentamycin sulfate
107 - 121°
C21H43N5O7·H2SO4
15, 4427
575.67

Chemical Identifiers

1405-41-0
C21H43N5O7·H2SO4
MFCD00270181
Garamycin, Gentamicin Sulfate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May damage fertility. May damage the unborn child.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

GHS P Statement:
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF exposed or concerned: Get medical advice/attention.
Obtain special instructions before use.
In case of inadequate ventilation wear respiratory protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only