missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Gentamicin sulfate

Gentamicin sulfate, CAS # 1405-41-0, is an aminoglycoside mainly used to inhibit the growth of Gram-negative bacteria.

Supplier:  Thermo Scientific Chemicals 455310010

 View more versions of this product

Catalog No. AC455310010


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Gentamicin sulfate is a bactericidal aminoglycoside antibiotic which shows activity against Gram-negative bacteria
  • It works by inhibiting protein biosynthesis through binding the bacterial ribosome 30S subunit

Applications

  • Gentamicin sulfate has been used in studies to determine wound healing due to its antibiotic and antifungal properties
  • It has laboratory research applications in the study of a number of clinical conditions, including actinomycotic infection, appendicitis, cystic fibrosis, osteomyelitis, pelvic inflammatory disease, plague, gonorrhea, and tularemia
Specifications

Specifications

Gentamicin Sulfate
1405-41-0
Authentic
1 g
Solubility in water: soluble.
Gentamicin sulfate
218.0°C to 237.0°C
15% max.
Garamycin, Gentamicin Sulfate
≥590 IU/mg (Gentamycin)

Chemical Identifiers

1405-41-0
Garamycin, Gentamicin Sulfate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

May cause allergy or asthma symptoms or breathing difficulties if inhaled, May cause an allergic skin reaction

Respiratory sensitisation (category 1), Skin sensitisation (category 1)

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only