missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amsbio LLC (formerly Genlantis) Subculturing Reagents
Functionally tested subculturing reagents for primary cells
$129.92 - $391.27
Specifications
| Form | Liquid |
|---|---|
| Quantity | 100 mL |
Includes: PR090100K: Hank's Buffered Saline Solution, 100mL; Trypsin/EDTA, 100mL; Trypsin Neutralizing Solution, 100mL
Specifications
| Liquid | |
| 100 mL |