Learn More
Genlantis™ phCMV Reporter Vectors DFS Item
High-level expression vectors for GFP or Luciferase fusion protein expression
Supplier:  Genlantis™ P013400
 View more versions of this product
Catalog No. P013400
Only null left
Explore available promotions
Description
Description
  • Modified hCMV immediate-early promoter provides high level constitutive expression pomoter in a wide variety of cells and cell types
  • Convenient choice of GFP or Luciferase reporter genes for control expression experiments
  • Two multiple cloning sites (MCS) for cloning gene of choice upstream or downstream of reporter gene (in fusion)
  • Kanamycin/Neomycin resistance gene provide efficient vector selection in E. coli (with Kanamycin sulfate) or mammalian cells (with Neomycin Sulfate)
  • SV40 polyadenylation sign provides for stable mRNA by providing efficient transcription termination and polyadenylation
  • PUC Origin of replication provides high copy number of vector in E. coli
Specifications
Specifications
Kanamycin/Neomycin
High
-20°C
20 μg
Used for either transient transfection or for generating stable cells lines expressing the reporter gene alone or in fusion with gene of interest
hCMV
Green Fluorescent Protein
0.5 μg/μL
phCMV N-GFP FSR vector
Product Suggestions
Product Suggestions