Please login to your online account to display your discounted pricing

Genlantis™ phCMV Reporter Vectors

High-level expression vectors for GFP or Luciferase fusion protein expression

$1,058.55 - $1,058.55

Specifications

Concentration 0.5μg/μL
Promoter hCMV
For Use With (Application) Used for either transient transfection or for generating stable cells lines expressing the reporter gene alone or in fusion with gene of interest
Marker Gene Kanamycin/Neomycin
Protein Expression Level High
View More Specs
Products
Catalog Number Mfr. No. Reporter Gene Vector Price Quantity    

P003400

 
genlantis
P003400
Green Fluorescent Protein phCMV C-GFP FSR vector Each for $1,058.55

P003500

 
genlantis
P003500
Luciferase phCMV-C-Luciferase FSR vector Each for $1,058.55

P013500

 
genlantis
P013500
Luciferase phCMV N-Luciferase vector Each for $1,058.55

P013400

 
genlantis
P013400
Green Fluorescent Protein phCMV N-GFP FSR vector Each for $1,058.55
Description & Specifications

Specifications

Concentration 0.5μg/μL
Promoter hCMV
For Use With (Application) Used for either transient transfection or for generating stable cells lines expressing the reporter gene alone or in fusion with gene of interest
Marker Gene Kanamycin/Neomycin
Protein Expression Level High
Quantity 20μg
Storage Requirements -20°C

  • Modified hCMV immediate-early promoter provides high level constitutive expression pomoter in a wide variety of cells and cell types
  • Convenient choice of GFP or Luciferase reporter genes for control expression experiments
  • Two multiple cloning sites (MCS) for cloning gene of choice upstream or downstream of reporter gene (in fusion)
  • Kanamycin/Neomycin resistance gene provide efficient vector selection in E. coli (with Kanamycin sulfate) or mammalian cells (with Neomycin Sulfate)
  • SV40 polyadenylation sign provides for stable mRNA by providing efficient transcription termination and polyadenylation
  • PUC Origin of replication provides high copy number of vector in E. coli

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.