missing translation for 'onlineSavingsMsg'
Learn More

Genlantis™ Genlantis™GenePORTER™ Gold Transfection Reagent

Catalog No. T204115
Encompass
Change view
Click to view available options
Quantity:
400 x 1.5 mL
800 x 1.5 mL
4000 x 15 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T204115 4000 x 15 mL
T204015 400 x 1.5 mL
T204030 800 x 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T204115 Supplier Genlantis™ Supplier No. T204115
Only null left

GenePORTER™ Gold Transfection Reagent is our newest, highly effective and reproducible lipid based gene delivery formulation for the widest variety of cell lines, especially the easy-to-transfect cell lines. It utilizes our Advanced Carrier Enhancement (ACE) technology that maximizes transfection efficiency and transgene expression levels with minimal cytotoxicity. The GenePORTER Gold Reagent and protocol have been optimized to provide the fastest, easiest, and most cost effective transfection reactions possible. Because of its simplicity and cost advantage, the GenePORTER Gold Transfection Reagent is ideal for high throughput or large scale applications where the need for cost control and great results are highly desired.

  • Super fast protocol, optimized for same-day transfection
  • Effective in a broad range of cell lines
  • Ideal for high-throughput applications
  • Value priced - your research dollars will go further
  • Unique Advanced Conjugation Enhancement (ACE) technology promotes effective delivery with exceptionally low cytotoxicity

Specifications

Includes 1 x 15mL GenePorter™ Gold Reagent, 1 x 100mL GP Gold Diluent, 1 x 10mL GP Gold Enhancer
Content And Storage 4°C
For Use With (Application) Ideal for high throughput applications, for same-day transfection in a broad range of cell lines
Quantity 4000 x 15 mL