missing translation for 'onlineSavingsMsg'
Learn More

Genistein 98.0+%, TCI America™

Catalog No. p-7130217
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0272100MG 100 mg
G02721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G0272100MG Supplier TCI America Supplier No. G0272100MG
Only null left

Chemical Identifiers

CAS 446-72-0
Molecular Formula C15H10O5
Molecular Weight (g/mol) 270.24
MDL Number MFCD00016952
InChI Key TZBJGXHYKVUXJN-UHFFFAOYSA-N
Synonym genistein, prunetol, genisteol, 4',5,7-trihydroxyisoflavone, genisterin, sophoricol, 5,7,4'-trihydroxyisoflavone, bonistein, 5,7-dihydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, genestein
PubChem CID 5280961
ChEBI CHEBI:28088
IUPAC Name 5,7-dihydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one
SMILES OC1=CC=C(C=C1)C1=COC2=CC(O)=CC(O)=C2C1=O

Specifications

Quantity 100 mg
Formula Weight 270.24
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Genistein
RTECSNumber NR2392000
TSCA Yes
Recommended Storage Refrigerator