missing translation for 'onlineSavingsMsg'
Learn More

Genistein, MP Biomedicals™

Catalog No. p-4404910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15235510 10 mg
ICN15235550 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15235510 Supplier MP Biomedicals Inc Supplier No. 0215235510

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 446-72-0
Molecular Formula C15H10O5
Molecular Weight (g/mol) 270.24
MDL Number MFCD00016952
InChI Key TZBJGXHYKVUXJN-UHFFFAOYSA-N
Synonym genistein, prunetol, genisteol, 4',5,7-trihydroxyisoflavone, genisterin, sophoricol, 5,7,4'-trihydroxyisoflavone, bonistein, 5,7-dihydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, genestein
PubChem CID 5280961
ChEBI CHEBI:28088
IUPAC Name 5,7-dihydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one
SMILES OC1=CC=C(C=C1)C1=COC2=CC(O)=CC(O)=C2C1=O

Specifications

Color White
Quantity 10 mg
Formula Weight 270.2
Physical Form Free-flowing Powder
Chemical Name or Material Genistein