missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Genetic Traits Taste Testing Strips
Demonstrate how the ability to taste a certain compound is based on heredity with these Genetic Traits Taste Testing Strips.
$7.46 - $17.07
Specifications
| Grade | 7 to 12 |
|---|---|
| Includes | 1 Vial of 100 Test Strips |
| Material | Paper |
| Class Size | 100 Students |
| Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
|
S85286A
|
Eisco™
FSC1034 |
Control Paper Taste Strip |
Pack of 100 Each for $7.46
|
|
|||||
|
S85287A
|
Eisco™
FSC1031 |
PTC Paper Taste Strip |
Pack of 100 Each for $15.07
|
|
|||||
|
S85288A
|
Eisco™
FSC1032 |
Sodium Benzoate Paper Taste Strip |
Pack of 100 Each for $15.07
|
|
|||||
|
S85289A
|
Eisco™
FSC1033 |
Thiourea Paper Taste Strip |
Pack of 100 Each for $17.07
|
|
|||||
Description
Is it sweet, salty, bitter or tasteless?
- Depending on their own genetics, students will experience different tastes from the test strips
- PTC — seven out of 10 students will detect a definite taste; remainder will not
- Sodium benzoate — genetics determine if the paper tastes sweet, salty, bitter or tasteless If a student can taste PTC, he may not be able to taste thiourea
- Control strips have no PTC, sodium benzoate or thiourea
Order Info
Each test strip comes in a pack of 100.
Specifications
| 7 to 12 | |
| 1 Vial of 100 Test Strips | |
| Paper | |
| 100 Students |