Learn More
Eisco™ Genetic Traits Taste Testing Strips
Demonstrate how the ability to taste a certain compound is based on heredity with these Genetic Traits Taste Testing Strips.
Supplier:  Eisco™ FSC1032
 View more versions of this product
Catalog No. S85288A
Only null left
Description
Description
Is it sweet, salty, bitter or tasteless?
  • Depending on their own genetics, students will experience different tastes from the test strips
  • PTC — seven out of 10 students will detect a definite taste; remainder will not
  • Sodium benzoate — genetics determine if the paper tastes sweet, salty, bitter or tasteless If a student can taste PTC, he may not be able to taste thiourea
  • Control strips have no PTC, sodium benzoate or thiourea
Order Info
Each test strip comes in a pack of 100.
Specifications
Chemical Identifiers
7 to 12
1 Vial of 100 Test Strips
Paper
100 Students
Specifications
7 to 12
1 Vial of 100 Test Strips
100 Students
Genetics determine if the paper tastes sweet, salty, bitter or tasteless if a student can taste ptc, he may not be able to taste thiourea
Paper
Sodium Benzoate Paper Taste Strip
Product Suggestions
Product Suggestions