missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Genetic Traits Taste Testing Strips

Catalog No. S85286A
Change view
Click to view available options
Product Type:
Control Paper Taste Strip
PTC Paper Taste Strip
Sodium Benzoate Paper Taste Strip
Thiourea Paper Taste Strip
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S85286A Control Paper Taste Strip
S85287A PTC Paper Taste Strip
S85288A Sodium Benzoate Paper Taste Strip
S85289A Thiourea Paper Taste Strip
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S85286A Supplier Eisco™ Supplier No. FSC1034

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Demonstrate how the ability to taste a certain compound is based on heredity with these Genetic Traits Taste Testing Strips.

Is it sweet, salty, bitter or tasteless?

  • Depending on their own genetics, students will experience different tastes from the test strips
  • PTC — seven out of 10 students will detect a definite taste; remainder will not
  • Sodium benzoate — genetics determine if the paper tastes sweet, salty, bitter or tasteless If a student can taste PTC, he may not be able to taste thiourea
  • Control strips have no PTC, sodium benzoate or thiourea

Order Info

Each test strip comes in a pack of 100.

Specifications

Grade 7 to 12
For Use With (Application) Used to demonstrates the effects of heredity on taste
Includes 1 Vial of 100 Test Strips
Material Paper
Class Size 100 Students
Product Type Control Paper Taste Strip