missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Geneclean™ SPIN Glassmilk™

Geneclean™ SPIN Glassmilk™ is a prepared aqueous suspension of proprietary silica matrix that contains TBE Modifier so it can be used with TAE- or TBE-buffered agarose gels.

$159.46 - $865.27

Specifications

Product Type Kits and Assays
Content And Storage Store at 2°C to 8°C
For Use With (Application) Nucleic Acid isolation and purification
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MP111101201
View Documents Promotion Details
MP Biomedicals™
111101201
30 mL
Each for $159.46
Only null left
 
MP111101601
View Documents Promotion Details
MP Biomedicals™
111101601
180 mL
Each for $865.27
Only null left
 
Description

Description

  • GeneClean SPIN Glassmilk is a specially prepared aqueous suspension of proprietary silica matrix.
  • It contains a built in TBE Modifier so it can be used with TAE- or TBE-buffered agarose gels.
  • It is used to bind DNA during purification procedures that utilize GeneClean Technology.
  • Aqueous Solution
Specifications

Specifications

Kits and Assays
Store at 2°C to 8°C
Nucleic Acid isolation and purification
Videos
SDS
Documents

Documents

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.