missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Geneclean™ SPIN Glassmilk™

Catalog No. MP111101201
Change view
Click to view available options
Quantity:
30 mL
180 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP111101201 30 mL
MP111101601 180 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP111101201 Supplier MP Biomedicals™ Supplier No. 111101201
Only null left

Geneclean™ SPIN Glassmilk™ is a prepared aqueous suspension of proprietary silica matrix that contains TBE Modifier so it can be used with TAE- or TBE-buffered agarose gels.

  • GeneClean SPIN Glassmilk is a specially prepared aqueous suspension of proprietary silica matrix.
  • It contains a built in TBE Modifier so it can be used with TAE- or TBE-buffered agarose gels.
  • It is used to bind DNA during purification procedures that utilize GeneClean Technology.
  • Aqueous Solution

Specifications

Product Type Kits and Assays
Content And Storage Store at 2°C to 8°C
For Use With (Application) Nucleic Acid isolation and purification
Quantity 30 mL

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.