missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GeneArt™ Gibson Assembly HiFi Master Mix

Catalog No. a46627
Encompass
Change view
Click to view available options
Quantity:
10 Reactions
200 Reactions
50 Reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A46627 10 Reactions
A46628 50 Reactions
A46629 200 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A46627 Supplier Invitrogen™ Supplier No. A46627
Only null left

GeneArt Gibson Assembly HiFi Master Mix enables DNA assembly and cloning via a technique that allows multiple overlapping DNA fragments to be seamlessly linked in a one-step, 15–60 minute isothermal reaction.

GeneArt Gibson Assembly HiFi Master Mix enables seamless, one-step, single-tube assembly of multiple overlapping DNA fragments in an isothermal reaction. This flexible DNA assembly methodology works for both small and large constructs, accommodating single or multiple inserts. PCR amplified fragments can be assembled without purification and transformed following a rapid competent cell transformation protocol. This approach significantly reduces hands-on time without great losses in positive clone yields. The resulting products are suitable for various downstream applications, including competent cell transformation, PCR, rolling circle amplification (RCA), synthetic biology, in vitro cell-free workflows, and mutagenesis.

Features of the GeneArt Gibson Assembly HiFi Master Mix include:

  • Simple—seamlessly assemble and clone up to six DNA fragments in a single reaction
  • Efficient—high fidelity provides more correct clones than other methods
  • Flexible—design guidelines allow assembly into any vector of your choice
  • Convenient—available as master mix kits and cloning kits complete with chemical or electrocompetent cells
  • Trusted—over 4000 citations in the scientific literature highlighting great success

Seamless assembly of multiple fragments

GeneArt Gibson Assembly HiFi cloning is a simple, one-step process whereby up to six fragments are combined in a proprietary enzymatic mix in order to assemble DNA fragments with shared terminal end homology without leaving any extra sequences or scars behind (seamless). After a 15–60 minute incubation, a portion of the assembly reaction is then transformed into chemically competent or electrocompetent cells, yielding clones that are ready for analysis the next day. The required 20- to 40-base pair end homology is designed into the de novo fragment for synthesis or can be easily engineered by PCR amplification with custom DNA oligos. This special enzyme mix creates a seamless and covalently bound DNA construct providing high efficiency.

Robust method for maximum efficiency

Due to the covalently bound final product, the GeneArt Gibson Assembly HiFi method allows the utilization of chemically competent cells or electrocompetent cells for the highest transformation efficiency, improving the success of finding the right clone, particularly for more challenging constructs. There is no need for restriction enzymes, ligation, or recombination sites, and the method provides a perfectly seamless construct without unwanted extra bases.

Provides great versatility

The GeneArt Gibson Assembly HiFi method provides versatility, can streamline many techniques through the rapid combination, addition, deletion, or exchange of DNA segments, and eliminates the need to subclone, saving time and effort in the cloning workflow.

Convenient formats

GeneArt Gibson Assembly HiFi kits are available in multiple formats, including:

  • GeneArt Gibson Assembly HiFi Cloning Kit, chemically competent cells (Part No. A46624 with One Shot TOP10 chemically competent cells)
  • GeneArt Gibson Assembly HiFi Cloning Kit, electrocompetent cells (Part No. A46626 with ElectroMAX DH10B electrocompetent cells)
  • GeneArt Gibson Assembly HiFi Master Mix kits (cloning kit without the cells) (this page)

Specifications

Cloning Method Seamless cloning
Content And Storage Includes master mix, positive control, and water and accommodates the use of your own competent cells.

Control Type Positive Control
For Use With (Application) Seamless Cloning & Genetic Assembly
No. of Reactions 10 Reactions
Number of Fragments Up to 6 fragments
Product Line GENEART
Product Type HiFi Master Mix
Quantity 10 Reactions
Shipping Condition Ambient, Wet Ice, Dry Ice
Workflow Step DNA Assembly
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.