missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GeneArt™ Gibson Assembly EX Cloning Kit, chemically competent cells

Catalog No. A46633
Encompass
Change view
Click to view available options
Quantity:
10 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A46633 10 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A46633 Supplier Invitrogen™ Supplier No. A46633

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The GeneArt Gibson Assembly EX Cloning Kit enables DNA assembly via a technique that allows up to 15 overlapping DNA fragments to be seamlessly linked in a single-tube, two-step optimized reaction.

The GeneArt Gibson Assembly EX Cloning Kit enables DNA assembly via a technique that allows up to 15 overlapping DNA fragments to be seamlessly linked in a single-tube, two-step optimized reaction. DNA fragments of different lengths are uniformly assembled using complementary overlaps between fragments. The inherent flexibility and success of this approach is suitable for small and large DNA constructs, includes both single and multiple inserts, and has been used to build entire genomes. The resulting products can be used for a variety of downstream applications, including transformation, PCR, and rolling circle amplification (RCA). The GeneArt Gibson Assembly EX Cloning Kit, chemically competent cells, is a complete kit that includes master mix, positive control, water, and One Shot TOP10 chemically competent E. coli.

Features of the GeneArt Gibson Assembly EX Cloning Kit include:

  • Simple—seamlessly assemble and clone up to 15 DNA fragments in a single reaction
  • Efficient—robust efficiency provides successful clones for both simple and challenging constructs
  • Flexible—design guidelines allow assembly into any vector of your choice
  • Convenient—available as master mix kits and cloning kits complete with chemical or electrocompetent cells
  • Trusted—over 4000 citations in the scientific literature highlighting great success

Seamless assembly of up to 15 fragments

GeneArt Gibson Assembly EX cloning is a robust, single-tube, two-step process whereby up to 15 inserts and vector are combined in a proprietary enzymatic mix in order to assemble DNA fragments with shared terminal end homology without leaving any extra sequences or scars behind (seamless). After this dually optimized reaction is complete, a portion of the assembly reaction is then transformed into chemically competent or electrocompetent cells, yielding clones ready for analysis the next day. The required 20- to 40-base pair end homology is designed into the de novo fragment for synthesis or can be easily engineered by PCR amplification with custom DNA oligos. This special enzyme mix creates a seamless and covalently bound DNA construct providing high efficiency.

Robust method for maximum efficiency

Due to the covalently bound final product, the GeneArt Gibson Assembly EX method allows the utilization of chemically competent cells or electrocompetent cells for the highest transformation efficiency, especially important for challenging constructs. There is no need for restriction enzymes, ligation, or recombination sites, and the method provides a perfectly seamless construct without unwanted extra bases.

Provides great versatility

The GeneArt Gibson Assembly EX method provides versatility, can streamline many techniques through the rapid combination, addition, deletion, or exchange of DNA segments, and eliminates the need to subclone, saving time and effort in the cloning workflow.

Specifications

Bacterial or Yeast Strain TOP10
Cell Type Chemically Competent E. coli
Cloning Method Seamless cloning
Content And Storage Includes master mix, positive control, water, and One Shot TOP10 chemically competent E. coli.

Control Type Positive Control
Format Kit
For Use With (Application) Seamless Cloning & Genetic Assembly
Number of Fragments Up to 15 fragments
Product Line GENEART
Product Type Cloning Kit
Quantity 10 Reactions
Shipping Condition Dry Ice
Workflow Step DNA Assembly
Show More Show Less
How big do DNA fragment homology arms need to be when using Gibson Assembly kits?

For fragments that are 8 kb or lesser in length, an overlap of 20-40 bp is recommended. For fragments greater than 8 kb in length, an overlap of 40-100 bp is recommended.

What type of vector can I use with Gibson Assembly kits?

Any linearized vector can be used with Gibson Assembly kits. We recommend using high-copy vectors for small inserts (<10 kb) and low-copy vectors for large inserts or toxic genes.

What type of inserts can I use with Gibson Assembly kits?

Any linearized DNA fragment, including PCR products, restriction digestion products, and GeneArt Strings can be used with Gibson Assembly kits.

What is the difference between the Gibson Assembly HiFi and the Gibson Assembly EX kits?

Gibson Assembly HiFi kits provide the quickest and easiest way to assemble up to six fragments in a single-tube reaction. The more robust Gibson Assembly EX kits enable assembly of up to 15 fragments in a single-tube, two-step reaction.

For Research Use Only. Not for use in diagnostic procedures.