missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Gemifloxacin mesylate

Catalog No. AC465972500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465972500 250 mg
AC465970010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465972500 Supplier Thermo Scientific Chemicals Supplier No. 465972500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 210353-53-0 | C19H24FN5O7S, C18 H20 F N5 O4 . C H4 O3 S | 485.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 210353-53-0
Molecular Formula C18 H20 F N5 O4 . C H4 O3 S

Specifications

Quantity 250 mg
Formula Weight 485.49
Physical Form Powder
Product Type Reagent
Purity ≥97.5% (HPLC)
Chemical Name or Material Gemifloxacin mesylate

RUO – Research Use Only