missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gemcitabine Hydrochloride 98.0+%, TCI America™
$206.90 - $973.03
Chemical Identifiers
| CAS | 122111-03-9 |
|---|---|
| Molecular Formula | C9H12ClF2N3O4 |
| Molecular Weight (g/mol) | 299.659 |
| MDL Number | MFCD01735988 |
| InChI Key | OKKDEIYWILRZIA-OSZBKLCCSA-N |
| Synonym | gemcitabine hcl, 2',2'-difluorodeoxycytidine monohydrochloride, 2'-deoxy-2',2'-difluorocytidine hydrochloride, 2 1h-pyrimidinone, 4-amino-1-2-deoxy-2,2-difluoro-d-erythro-pentofuranosyl |
| PubChem CID | 44558863 |
| ChEBI | CHEBI:31647 |
| IUPAC Name | 4-amino-1-[(2R,4R,5R)-3,3-difluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one;hydron;chloride |
| SMILES | [H+].C1=CN(C(=O)N=C1N)C2C(C(C(O2)CO)O)(F)F.[Cl-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
G0367100MG
|
TCI America
G0367100MG |
100 mg |
Each for $206.90
|
|
|||||
|
G03671G
|
TCI America
G03671G |
1 g |
Each for $973.03
|
|
|||||
Chemical Identifiers
| 122111-03-9 | |
| 299.659 | |
| OKKDEIYWILRZIA-OSZBKLCCSA-N | |
| 44558863 | |
| 4-amino-1-[(2R,4R,5R)-3,3-difluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one;hydron;chloride |
| C9H12ClF2N3O4 | |
| MFCD01735988 | |
| gemcitabine hcl, 2',2'-difluorodeoxycytidine monohydrochloride, 2'-deoxy-2',2'-difluorocytidine hydrochloride, 2 1h-pyrimidinone, 4-amino-1-2-deoxy-2,2-difluoro-d-erythro-pentofuranosyl | |
| CHEBI:31647 | |
| [H+].C1=CN(C(=O)N=C1N)C2C(C(C(O2)CO)O)(F)F.[Cl-] |
Specifications
| 122111-03-9 | |
| White | |
| MFCD01735988 | |
| gemcitabine hcl, 2',2'-difluorodeoxycytidine monohydrochloride, 2'-deoxy-2',2'-difluorocytidine hydrochloride, 2 1h-pyrimidinone, 4-amino-1-2-deoxy-2,2-difluoro-d-erythro-pentofuranosyl | |
| [H+].C1=CN(C(=O)N=C1N)C2C(C(C(O2)CO)O)(F)F.[Cl-] | |
| 299.659 | |
| CHEBI:31647 | |
| ≥98.0% (HPLC,N) | |
| Gemcitabine Hydrochloride |
| 292°C | |
| C9H12ClF2N3O4 | |
| 100 mg | |
| OKKDEIYWILRZIA-OSZBKLCCSA-N | |
| 4-amino-1-[(2R,4R,5R)-3,3-difluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one;hydron;chloride | |
| 44558863 | |
| 299.66 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : HA3840000
TSCA : No