missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gemcitabine Hydrochloride 98.0+%, TCI America™
Supplier: TCI America G03671G
Specifications
| Gemcitabine Hydrochloride | |
| 292°C | |
| C9H12ClF2N3O4 | |
| 1 g | |
| OKKDEIYWILRZIA-OSZBKLCCSA-N | |
| 4-amino-1-[(2R,4R,5R)-3,3-difluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one;hydron;chloride | |
| 44558863 | |
| 299.66 | |
| Crystalline Powder |
| 122111-03-9 | |
| White | |
| MFCD01735988 | |
| gemcitabine hcl, 2',2'-difluorodeoxycytidine monohydrochloride, 2'-deoxy-2',2'-difluorocytidine hydrochloride, 2 1h-pyrimidinone, 4-amino-1-2-deoxy-2,2-difluoro-d-erythro-pentofuranosyl | |
| [H+].C1=CN(C(=O)N=C1N)C2C(C(C(O2)CO)O)(F)F.[Cl-] | |
| 299.659 | |
| CHEBI:31647 | |
| ≥98.0% (HPLC,N) |
Chemical Identifiers
| 122111-03-9 | |
| 299.659 | |
| OKKDEIYWILRZIA-OSZBKLCCSA-N | |
| 44558863 | |
| 4-amino-1-[(2R,4R,5R)-3,3-difluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one;hydron;chloride |
| C9H12ClF2N3O4 | |
| MFCD01735988 | |
| gemcitabine hcl, 2',2'-difluorodeoxycytidine monohydrochloride, 2'-deoxy-2',2'-difluorocytidine hydrochloride, 2 1h-pyrimidinone, 4-amino-1-2-deoxy-2,2-difluoro-d-erythro-pentofuranosyl | |
| CHEBI:31647 | |
| [H+].C1=CN(C(=O)N=C1N)C2C(C(C(O2)CO)O)(F)F.[Cl-] |
Safety and Handling
RTECSNumber : HA3840000
TSCA : No