missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Gefitinib-based PROTAC™ 3
Description
Gefitinib-based PROTAC® 3 is a potent EGFR PROTAC™ Degrader. Comprises an EGFR inhibitor gefitinib (Iressa Cat.No. 3000) conjugated to a VHL ligand via a linker. Induces EGFR degradation (DC50 values are 11.7 nM and 22.3 nM in HCC827 (exon 19 del) and H3255 (L858R mutation) cells, respectively). Exhibits no degradation of wild-type EGFR at concentrations up to 10 μM.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | (2 S,4 R)-1-(-2-(3-(2-((5-((4-((3-Chloro-4-fluorophenyl)amino)-7-methoxyquinazolin-6-yl)oxy)pentyl)oxy)ethoxy)propanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide |
| CAS | 2230821-27-7 |
| Quantity | 5 mg |
| Target | Active Degraders |
| Molecular Formula | C47H57ClFN7O8S |
| Purity | 0.98 |