missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GDNF Monoclonal Antibody (K92005_4F10), Invitrogen™
Mouse Monoclonal Antibody
$1,461.54 - $2,194.06
Specifications
| Antigen | GDNF |
|---|---|
| Clone | K92005_4F10 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Sequence of this protein is as follows: RNRQAAAANP ENSRGKGRRG QRGKNRGCVL TAIHLNVTDL GLGYETKEEL IFRYCSGSCD AAETTYDKIL KNLSRNRRLV
GDNF Monoclonal antibody specifically detects GDNF in Human samples. It is validated for Western BlotSpecifications
| GDNF | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| AI385739; Astrocyte-derived trophic factor; ATF; ATF1; ATF2; Gdnf; glial cell derived neurotrophic factor; glial cell line derived neurotrophic factor; Glial cell line derived neutrophic factor; glial cell line-derived neurotrophic factor; gndf; HFB1-GDNF; hGDNF; H-GDNF; HSCR3; mGDNF; neurotrophic factor | |
| Recombinant human GDNF (immunogen range is 91-170). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| GDNF |
| K92005_4F10 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P39905 | |
| 2668 | |
| IgG1 | |
| Protein A | |
| Antibody |